Mouse Recombinant CD274 (from HEK293 cells)
Supplier: Sino Biological
50010-M02H-100 50010-M02H-1
103664-372EA 179.78 USD
103664-372 103664-374 76642-096
Mouse Recombinant CD274 (from HEK293 cells)
Proteins and Peptides
A DNA sequence encoding the mouse Cd274 (NP_068693.1) (Met1-His239) was expressed with the Fc region of human IgG1 at the C-terminus.