Listeria monocytogenes Recombinant flagellin/flaA Protein (His Tag) (from E. coli)
Supplier: Sino Biological
40067-V07E-20 40067-V07E-100
103615-164EA 139.38 USD
103615-164 103663-600
Listeria monocytogenes Recombinant flagellin/flaA Protein (His Tag) (from E. coli)
Proteins and Peptides
A DNA sequence encoding the flaA (YP_013330.1) (Met1-Ser287) was expressed with a N-terminal polyhistidine tag.