Human Recombinant SerpinF1/PEDF Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
11104-H08H-50 11104-H08H-1
103680-586EA 116.1 USD
103680-586 103680-588 76641-788
Human Recombinant SerpinF1/PEDF Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human SERPINF1 (NP_002606.3) (Met 1-Pro 418) was fused with a polyhistidine tag at the C-terminus.