Human Recombinant Neuropilin-2/NRP2 Protein (His Tag) (from HEK293 Cells)
Supplier: Sino Biological
10695-H08H-50 10695-H08H-1
103679-740EA 148.35 USD
103679-740 103679-742 76641-140
Human Recombinant Neuropilin-2/NRP2 Protein (His Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the extracellular domain (Met 1-Tyr 855) ofhuman Neurophilin2 isoform 2 (NP_003863.2) was expressed with a C-terminal polyhistidine tag.