Human Recombinant ACVR2B/ActivinR-IIB Protein (ECD, Fc Tag) (from HEK293 Cells)
Supplier: Sino Biological
10229-H02H-100 10229-H02H-1
103620-884EA 300.98 USD
103620-884 103620-886 76641-502
Human Recombinant ACVR2B/ActivinR-IIB Protein (ECD, Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the N-terminal segment (Met 1-Thr 134) from the extracellular domain of human ACVR2B (NP_001097.2) was expressed with the fused human IgG1 Fc region at the C-terminus.