Human Recombinant TLR4/CD284 Protein (His Tag) (from Baculovirus Insect cells)
Supplier: Sino Biological
10146-H08B-100 10146-H08B-1
103620-856EA 300.98 USD
103620-856 103678-420 76640-874
Human Recombinant TLR4/CD284 Protein (His Tag) (from Baculovirus Insect cells)
Proteins and Peptides
A DNA sequence encoding the human TLR4 (Met1-Lys631) (O00206-1) was expressed with a C-terminal polyhistidine tag.