PPM-18 ≥95%

Supplier: Cayman Chemical Company

Synonyms: NSC 73233

13327-1 13327-5 13327-10 13327-25
76293-088EA 23.57 USD
76293-088 76293-090 76293-092 76293-094
PPM-18 ≥95%
PPM-18

  • Inhibits activation of NF-κB


An inhibitor NF-κB activation in vitro and in vivo (IC50 = 5 µM)

Storage Temperature: Freezer
CAS Number: 65240-86-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4ad0ea0de92022ef1a590ac0a65e4dcd44ce469c&vid=4ad0ea0de92022ef1a590ac0a65e4dcd44ce469c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d49712a091fb7567a8ea045a63f361467db4de6&vid=6d49712a091fb7567a8ea045a63f361467db4de6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥95%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR