SC-514 ≥98%

Supplier: Cayman Chemical Company
Danger

Synonyms: 4-amino-[2,3'-bithiophene]-5-carboxamide, IKK-2 Inhibitor, SC-514

10010267-5 10010267-10 10010267-25 10010267-50
76293-060EA 48.52 USD
76293-060 76293-062 76293-064 76293-066
SC-514 ≥98%
SC-514

  • Selective inhibitor of IKK2


A selective and reversible inhibitor of IKK2 (IC50 = 3-12 μM) that displays >10-fold selectivity over 28 other kinases; attenuates NF-κB-mediated gene expression in synovial fibroblasts, smooth muscle cells, and astrocytes

Formula: C₉H₈N₂OS₂
Storage Temperature: Freezer
MDL Number: MFCD00206459
CAS Number: 354812-17-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a199494492ff9b45fa6f0969d0c8750c0a656e9&vid=ca11e83e690b47d0f92510810c22913d677afac0&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=996c7bcb6cf8a40dcd360dde5dc6a1f9a28c0c61&vid=996c7bcb6cf8a40dcd360dde5dc6a1f9a28c0c61&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=75bea92ab323bdc9a9f9aff06fddfc77a6a9a1f8&vid=75bea92ab323bdc9a9f9aff06fddfc77a6a9a1f8&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa22b6d8a83dcb1882a5c655abb840b445a20b2b&vid=fa22b6d8a83dcb1882a5c655abb840b445a20b2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Formulation A crystalline solid
Purity ≥98%
Stability ≥ 2 years


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR