5-Bromo-2'-deoxyuridine (broxuridine)

Supplier: Anaspec Inc

Synonyms: 5-Bromo-2'-deoxyuridine5-BrdU5-Bromo-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione

AS-83224
103011-146EA 119.85 USD
103011-146
5-Bromo-2'-deoxyuridine (broxuridine)
5-Bromo-2'-deoxyuridine (Broxuridine)

Biomarker for cell cycle and cell proliferation


  • Spectral properties: Abs/Em = ND/none nm
  • Solvent system: DMSO
  • Form: Solid,
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice

Formula: C₉H₁₁BrN₂O₅
MW: 307.10 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00006529
CAS Number: 59-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR