Biocytin
Supplier: Anaspec Inc
Synonyms:
ε-Biotinoyl-L-lysine
, H-Lys(biotinyl)-OH
AS-60638
103003-288EA
190
USD
103003-288
Biocytin
Biocytin
More polar than biotin; used for cell tracing
- Spectral properties: Abs/Em = NA/NA nm
- Solvent system: water or DMSO
- Storage Temperature: -20°C desiccated and protected from light
- Shipping condition: RT
Formula:
C₁₆H₂₈N₄O₄S MW: 372.484 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00077319 CAS Number: 576-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...