Anti-His Tag Human Monoclonal Antibody [clone: 3D5]

Supplier: Enzo Life Sciences
ABS539-0200
76285-070EA 633 USD
76285-070
Anti-His Tag Human Monoclonal Antibody [clone: 3D5]
Antibodies
Recombinant monclonal antibody (Human IgG3κ).

Recommended Applications: ELISA, WB

Type: Primary
Antigen: His Tag
Clonality: Monoclonal
Clone: 3D5
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG3κ
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR