Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]

Supplier: Enzo Life Sciences
ABS459-0100
76284-786EA 652 USD
76284-786
Anti-MYC Mouse Monoclonal Antibody [clone: 9E10]
Antibodies
Recombinant monclonal antibody (Mouse Fab fragment κ).

Recommended Applications: IF, IHC (FS), IHC (PS), IP, WB

Type: Primary
Antigen: MYC
Clonality: Monoclonal
Clone: 9E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Fab fragment κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR