Anti-IL-5 Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P55BT-25UG 500-P55BT-50UG
10771-322EA 354.54 USD
10771-322 10771-324 10771-326
Anti-IL-5 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Murine IL-5. Anti­Murine IL-5­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: IL-5
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Murine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR