Anti-MDC (CCL22) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P107BT-25UG 500-P107BT-50UG
10771-354EA 354.54 USD
10771-354 10771-356 10771-358
Anti-MDC (CCL22) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human MDC (CCL22). Anti­Human MDC (CCL22)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: MDC (CCL22)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR