Anti-FGF-basic Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P18BT-25UG 500-P18BT-50UG
10775-088EA 339.8 USD
10775-088 10775-090 10775-092
Anti-FGF-basic Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human FGF-basic. Anti­Human FGF-basic­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: FGF-basic
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR