Anti-beta-NGF Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P85BT-25UG 500-P85BT-50UG
10775-780EA 354.54 USD
10775-780 10775-782 10775-784
Anti-beta-NGF Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human β-NGF. Anti­Human β-NGF­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: β-NGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR