Anti-BCA-1 (CXCL13) Rabbit Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P141BT-25UG 500-P141BT-50UG
10776-360EA 358.56 USD
10776-360 10776-362 10776-364
Anti-BCA-1 (CXCL13) Rabbit Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human BCA-1 (CXCL13). Anti­Human BCA-1 (CXCL13)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: BCA-1 (CXCL13)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR