Anti-IL-6 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P73-50UG 500-P73-100UG 500-P73-1MG
10775-642EA 339.8 USD
10775-642 10775-644 10775-646
Anti-IL-6 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-6. Anti­Rat IL-6­specific antibody was purified by affinity chromatography employing an immobilized Rat IL-6 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR