Anti-Wnt-1 Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P250-50UG 500-P250-100UG 500-P250-1MG
10777-254EA 339.8 USD
10777-254 10777-256 10777-258
Anti-Wnt-1 Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human Wnt-1. Anti­Human Wnt-1­specific antibody was purified by affinity chromatography employing an immobilized Human Wnt-1 matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: Wnt-1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR