Anti-TGF-alpha Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P16-50UG 500-P16-100UG 500-P16-1MG
10775-058EA 339.8 USD
10775-058 10775-060 10775-062
Anti-TGF-alpha Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human TGF-α. Anti­Human TGF-α­specific antibody was purified by affinity chromatography employing an immobilized Human TGF-α matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: TGF-alpha
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR