Anti-KGF (FGF-7) Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P19-50UG 500-P19-100UG 500-P19-1MG
10775-094EA 339.8 USD
10775-094 10775-096 10775-098
Anti-KGF (FGF-7) Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human KGF (FGF-7). Anti­Human KGF (FGF-7)­specific antibody was purified by affinity chromatography employing an immobilized Human KGF (FGF-7) matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: KGF (FGF-7)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR