Anti-EGF Rabbit Polyclonal Antibody

Supplier: Peprotech
500-P45-50UG 500-P45-100UG 500-P45-1MG
10775-338EA 339.8 USD
10775-338 10775-340 10775-342
Anti-EGF Rabbit Polyclonal Antibody
Antibodies
Produced from sera of rabbits immunized with highly pure Recombinant Human EGF. Anti­Human EGF­specific antibody was purified by affinity chromatography employing an immobilized Human EGF matrix.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: EGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: Rabbit IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR