Anti-Eotaxin-2 (CCL24) Goat Polyclonal Antibody (Biotin)

Supplier: Peprotech
500-P103GBT-25UG 500-P103GBT-50UG
10775-980EA 368.12 USD
10775-980 10775-982
Anti-Eotaxin-2 (CCL24) Goat Polyclonal Antibody (Biotin)
Antibodies
Produced from sera of goats immunized with highly pure Recombinant Human Eotaxin-2 (CCL24). Anti­Human Eotaxin-2 (CCL24)­specific antibody was purified by affinity chromatography and then biotinylated.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Secondary
Antigen: Eotaxin-2 (CCL24)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: Goat IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR