AMP+ MaxSpec® Kit, Cayman Chemical
Supplier: Cayman Chemical Company
710000-15
710000-50
710000-100
76210-956EA
81.81
USD
76210-956
76210-958
76210-960
AMP+ MaxSpec® Kit, Cayman Chemical
Assays
Cellular Assays
A novel method to increase MS sensitivity for detection of lipids.
Cayman’s AMP+ MaxSpec® Kit contains a positively charged reagent (AMP+) that can be coupled quantitatively to free carboxylic acids through an amide bond linkage. The resulting derivatized product thus contains a positively charged group that shows improved electrospray ionization and consequently improved MS sensitivity. The derivatization reagent is stable to MS/MS fragmentation and thus isobaric derivatives undergo compound-specific fragmentation to give unique fragment ions.