Mouse Recombinant E-selectin (SELE)
Supplier: Bon Opus Biosciences
CB50-500UG CB50-50UG CB50-10UG CB50-1MG
102497-270EA 1741.38 USD
102497-270 102497-272 102509-638 102509-640
Mouse Recombinant E-selectin (SELE)
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human E-selectin is produced by our Mammalian expression system and the target gene encoding Trp22-Pro556 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.