Human Recombinant CD48 antigen
Supplier: Bon Opus Biosciences
CB90-50UG CB90-10UG CB90-1MG CB90-500UG
102497-342EA 283.23 USD
102497-342 102508-868 102508-870 102508-872
Human Recombinant CD48 antigen
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human SLAMF2 is produced by our Mammalian expression system and the target gene encoding Gln27-Ser220 is expressed with a Fc, 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.