Human Recombinant Intercellular Adhesion Molecule 1
Supplier: Bon Opus Biosciences
CX35-1MG CX35-500UG CX35-50UG CX35-10UG
102497-954EA 2487.69 USD
102497-954 102497-956 102497-958 102508-980
Human Recombinant Intercellular Adhesion Molecule 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Intercellular Adhesion Molecule 1 is produced by our Mammalian expression system and the target gene encoding Asn26-Glu480 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.