Human Recombinant Putative Polycomb group protein ASXL1
Supplier: Bon Opus Biosciences
CG54-500UG CG54-50UG CG54-10UG CG54-1MG
102499-172EA 1899.69 USD
102499-172 102499-174 102509-122 102509-124
Human Recombinant Putative Polycomb group protein ASXL1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Additional sex combs-like protein 1 is produced by our E.coli expression system and the target gene encoding Lys1477-Arg1541 is expressed with a GST tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.