Human Recombinant Protein S100-A4
Supplier: Bon Opus Biosciences
CR29-10UG CR29-1MG CR29-50UG CR29-500UG
103529-180EA 85.08 USD
103529-180 103529-370 103529-454 103529-496
Human Recombinant Protein S100-A4
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human S100A4 is produced by our E.coli expression system and the target gene encoding Met1-Lys101 is expressed with a 6His at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.