Human Recombinant CD265
Supplier: Bon Opus Biosciences
CK64-10UG CK64-50UG CK64-500UG CK64-1MG
102970-186EA 102.31 USD
102970-186 102970-312 102970-406 102970-500
Human Recombinant CD265
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Receptor Activator of NF-kappa-B is produced by our Mammalian expression system and the target gene encoding Ile30-Pro212 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.