Mouse Recombinant Semaphorin-4C
Supplier: Bon Opus Biosciences
CJ70-10UG CJ70-50UG CJ70-500UG CJ70-1MG
102944-738EA 170.15 USD
102944-738 102944-740 102944-742 102944-744
Mouse Recombinant Semaphorin-4C
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Mouse Semaphorin 4C is produced by our Mammalian expression system and the target gene encoding Ala21-Gly664 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.