Human Recombinant Cytokine receptor common subunit gamma
Supplier: Bon Opus Biosciences
CJ53-10UG CJ53-50UG CJ53-500UG CJ53-1MG
102944-602EA 102.31 USD
102944-602 102944-604 102944-606 102944-608
Human Recombinant Cytokine receptor common subunit gamma
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Cytokine receptor common subunit gamma is produced by our Mammalian expression system and the target gene encoding Leu23-Ala262 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.