Human Recombinant GDNFR-beta
Supplier: Bon Opus Biosciences
CJ30-10UG CJ30-50UG CJ30-500UG CJ30-1MG
102944-442EA 57.08 USD
102944-442 102944-444 102944-446 102944-448
Human Recombinant GDNFR-beta
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human GDNF Receptor alpha 2 is produced by our Mammalian expression system and the target gene encoding Ser22-Ser441 is expressed with a Fc, 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.