Human Recombinant PLA2G1B/Phospholipase A2/Group IB phospholipase A2/PLA2/PLA2A/PPLA2

Supplier: Bon Opus Biosciences

CJ15-10UG CJ15-50UG CJ15-500UG CJ15-1MG
102944-322EA 170.15 USD
102944-322 102944-324 102944-326 102944-328
Human Recombinant PLA2G1B/Phospholipase A2/Group IB phospholipase A2/PLA2/PLA2A/PPLA2
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human PLA2G1B is produced by our Mammalian expression system and the target gene encoding Ala23-Ser148 is expressed with a 6His tag at the C-terminus.


Accessories information: Accessories not included.


Delivery information: Shipped Via FedEx


Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.


Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR