Human Recombinant Interleukin-17 Receptor A
Supplier: Bon Opus Biosciences
CI53-10UG CI53-50UG CI53-500UG CI53-1MG
102943-882EA 57.08 USD
102943-882 102943-884 102943-886 102943-888
Human Recombinant Interleukin-17 Receptor A
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Interleukin-17 receptor A is produced by our Mammalian expression system and the target gene encoding Leu33-Trp320 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.