Human Recombinant UPF0556 protein C19orf10
Supplier: Bon Opus Biosciences
CG64-10UG CG64-1MG CG64-500UG CG64-50UG
102509-126EA 50.62 USD
102509-126 102509-128 102509-130 102509-132
Human Recombinant UPF0556 protein C19orf10
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human SF20 is produced by our E.coli expression system and the target gene encoding Ser33-Leu173 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.