Human Recombinant Thioredoxin-Dependent Peroxide Reductase Mitochondrial
Supplier: Bon Opus Biosciences
CG26-10UG CG26-1MG CG26-500UG CG26-50UG
102865-262EA 107.69 USD
102865-262 102865-264 102865-266 102865-268
Human Recombinant Thioredoxin-Dependent Peroxide Reductase Mitochondrial
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Peroxiredoxin-3 is produced by our E.coli expression system and the target gene encoding Pro63-Gln256 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.