Human Recombinant MHC Class I Polypeptide-Related Sequence A
Supplier: Bon Opus Biosciences
CG11-10UG CG11-1MG CG11-500UG CG11-50UG
102865-174EA 107.69 USD
102865-174 102865-176 102865-178 102865-180
Human Recombinant MHC Class I Polypeptide-Related Sequence A
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human MHC Class I-related Protein A is produced by our Mammalian expression system and the target gene encoding Ala23-Glu308 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.