Human Recombinant DnaJ Homolog Subfamily B Member 1
Supplier: Bon Opus Biosciences
CF72-10UG CF72-1MG CF72-500UG CF72-50UG
102864-982EA 107.69 USD
102864-982 102864-984 102864-986 102864-988
Human Recombinant DnaJ Homolog Subfamily B Member 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Heat Shock 40 kDa Protein is produced by our E.coli expression system and the target gene encoding Gly2-Ile340 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.