Human Recombinant SH2 Domain-Containing Protein 1A
Supplier: Bon Opus Biosciences
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
- Consistent Sizing options
- Datasheet Included
- High Purity, Low Endotoxin levels
Recombinant Human SH2D1A is produced by our E.coli expression system and the target gene encoding Met1-Pro128 is expressed with a 6His tag at the N-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...