Human Recombinant Signal Transducer and Activator of Transcription 3
Supplier: Bon Opus Biosciences
CF29-10UG CF29-1MG CF29-500UG CF29-50UG
102864-678EA 170.15 USD
102864-678 102864-680 102864-682 102864-684
Human Recombinant Signal Transducer and Activator of Transcription 3
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human STAT3 is produced by our E.coli expression system and the target gene encoding Met1-Asn175 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.