Human Recombinant Microtubule-Associated Proteins 1A/1B Light Chain 3A

Supplier: Bon Opus Biosciences

CE88-10UG CE88-1MG CE88-500UG CE88-50UG
102864-398EA 107.69 USD
102864-398 102864-400 102864-402 102864-404
Human Recombinant Microtubule-Associated Proteins 1A/1B Light Chain 3A
Proteins and Peptides

Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products


  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels


Recombinant Human MAP1LC3A is produced by our E.coli expression system and the target gene encoding Met1-Phe121 is expressed with a 6His tag at the C-terminus.


Accessories information: Accessories not included.


Delivery information: Shipped Via FedEx


Packaging: Shipped domestic on polar packs in a 9"x6"x7" box, or shipped internationally on Dry Ice in a 14" cube box.


Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.

The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR