Human Recombinant Baculoviral IAP Repeat-Containing Protein 5
Supplier: Bon Opus Biosciences
CE87-10UG CE87-1MG CE87-500UG CE87-50UG
102864-390EA 107.69 USD
102864-390 102864-392 102864-394 102864-396
Human Recombinant Baculoviral IAP Repeat-Containing Protein 5
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human BIRC5 is produced by our E.coli expression system and the target gene encoding Met1-Asp142 is expressed.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.