Human Recombinant Leucine-Rich Alpha-2-Glycoprotein
Supplier: Bon Opus Biosciences
CX80-10UG CX80-1MG CX80-500UG CX80-50UG
102498-226EA 107.69 USD
102498-226 102498-228 102498-230 102498-232
Human Recombinant Leucine-Rich Alpha-2-Glycoprotein
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human LRG1 is produced by our Mammalian expression system and the target gene encoding Val36-Gln347 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.