Human Recombinant Protocadherin-10
Supplier: Bon Opus Biosciences
CX63-10UG CX63-1MG CX63-500UG CX63-50UG
102498-128EA 90.46 USD
102498-128 102498-130 102498-132 102498-134
Human Recombinant Protocadherin-10
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Protocadherin-10 is produced by our Mammalian expression system and the target gene encoding Gln19-Thr715 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.