Human Recombinant Cadherin-6
Supplier: Bon Opus Biosciences
CX39-10UG CX39-1MG CX39-500UG CX39-50UG
102497-984EA 107.69 USD
102497-984 102497-986 102497-988 102497-990
Human Recombinant Cadherin-6
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Cadherin-6 is produced by our Mammalian expression system and the target gene encoding Thr19-Ala615 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.