Human Recombinant Lysosome-Associated Membrane Glycoprotein 1
Supplier: Bon Opus Biosciences
CX15-10UG CX15-1MG CX15-500UG CX15-50UG
102497-848EA 107.69 USD
102497-848 102497-850 102497-852 102497-854
Human Recombinant Lysosome-Associated Membrane Glycoprotein 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human LAMP1 is produced by our Mammalian expression system and the target gene encoding Ala29-Met382 is expressed with a Fc tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.