Human Recombinant Granulocyte-macrophage colony-stimulating factor is also known as Colony-stimulating factor
Supplier: Bon Opus Biosciences
CC79-10UG CC79-1MG CC79-500UG CC79-50UG
102508-932EA 107.69 USD
102508-932 102508-934 102508-936 102508-938
Human Recombinant Granulocyte-macrophage colony-stimulating factor is also known as Colony-stimulating factor
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Granulocyte-Macrophage Colony-Stimulating Factor is produced by our Mammalian expression system and the target gene encoding Ala18-Glu144 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.