Human Recombinant Regenerating islet-derived protein 3-gamma
Supplier: Bon Opus Biosciences
CC77-10UG CC77-1MG CC77-500UG CC77-50UG
102497-638EA 170.15 USD
102497-638 102497-640 102508-928 102508-930
Human Recombinant Regenerating islet-derived protein 3-gamma
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human Regenerating islet-derived protein 3-gamma is produced by our Mammalian expression system and the target gene encoding Glu27-Asp175 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.