Human Recombinant Cell Growth Regulator with EF Hand Domain Protein 1
Supplier: Bon Opus Biosciences
CA79-10UG CA79-1MG CA79-500UG CA79-50UG
102863-742EA 118.46 USD
102863-742 102863-744 102863-746 102863-748
Human Recombinant Cell Growth Regulator with EF Hand Domain Protein 1
Proteins and Peptides
Bon Opus Biosciences offers quality recombinant proteins products covering a broad collection of cytokines, enzymes, diagnostic and detection reagents, and other protein-related products
  • Consistent Sizing options
  • Datasheet Included
  • High Purity, Low Endotoxin levels
Recombinant Human CGREF1 is produced by our Mammalian expression system and the target gene encoding Ala20-Ile301 is expressed with a 6His tag at the C-terminus.
Accessories information: Accessories not included.
Delivery information: Shipped Via FedEx
Packaging: Shipped at ambient temperature via FedEx envelope.
Caution: This product list contains purified protein and other non-biological reagents, which are minimum risk biologicals, non-toxic and non-hazardous, and are intended for laboratory research use only.
The products are not intended for human or diagnostic use, and shall not be injected or ingested by any human.